Assay Details
| Gene Symbol: FGF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204888.1 | NP_990219.1 | 2 - 3 | 351 | 65 | |
| XM_015291369.2 | XP_015146855.1 | 2 - 3 | 459 | 65 | ||
| XM_015291371.2 | XP_015146857.1 | 3 - 4 | 327 | 65 | ||
| XM_025153311.1 | XP_025009079.1 | 2 - 3 | 128 | 65 | ||
| XM_025153312.1 | XP_025009080.1 | 2 - 3 | 161 | 65 | ||
| GenBank mRNA | AF108754.1 | - | 2 - 3 | 351 | 65 | |
| AF199602.1 | - | 3 - 4 | 271 | 65 | ||
| AF199603.1 | - | 1 - 2 | 299 | 65 | ||
More Information
Set Membership: |
|

FGF12
Genomic Map