Assay Details
| Gene Symbol: INS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205222.3 | NP_990553.1 | 2 - 3 | 297 | 102 | |
| XM_015286579.2 | XP_015142065.1 | 1 - 2 | 6898 | 102 | ||
| GenBank mRNA | BX278627.4 | - | 2 - 3 | 225 | 102 | |
| X58993.1 | - | 209 | 102 | |||
More Information
Set Membership: |
|

INS
Genomic Map