Assay Details
| Gene Symbol: RAB2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205228.1 | NP_990559.1 | 6 - 7 | 476 | 79 | |
| GenBank mRNA | X84220.1 | - | 476 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB2A| Gene Symbol: RAB2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205228.1 | NP_990559.1 | 6 - 7 | 476 | 79 | |
| GenBank mRNA | X84220.1 | - | 476 | 79 | ||
Set Membership: |
|