Assay Details
| Gene Symbol: MYLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205392.1 | NP_990723.1 | 6 - 7 | 1963 | 81 | |
| GenBank mRNA | M81787.1 | - | 6 - 7 | 1963 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYLK2| Gene Symbol: MYLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205392.1 | NP_990723.1 | 6 - 7 | 1963 | 81 | |
| GenBank mRNA | M81787.1 | - | 6 - 7 | 1963 | 81 | |
Set Membership: |
|