Assay Details
| Gene Symbol: SYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205394.1 | NP_990725.1 | 4 - 5 | 618 | 69 | |
| XM_015293015.1 | XP_015148501.1 | 5 - 6 | 766 | 69 | ||
| XM_015293016.2 | XP_015148502.1 | 4 - 5 | 709 | 69 | ||
| GenBank mRNA | U27708.1 | - | 4 - 5 | 618 | 69 | |
More Information
Set Membership: |
|

SYP
Genomic Map