Assay Details
| Gene Symbol: TGFBR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205428.1 | NP_990759.1 | 1 - 2 | 106 | 83 | |
| XM_015281321.2 | XP_015136807.1 | 1 - 2 | 302 | 83 | ||
| XM_015281322.2 | XP_015136808.1 | 1 - 2 | 305 | 83 | ||
| GenBank mRNA | L18784.1 | - | 1 - 2 | 106 | 83 | |
More Information
Set Membership: |
|

TGFBR2
Genomic Map