Assay Details
| Gene Symbol: LGALS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205495.1 | NP_990826.1 | 2 - 3 | 112 | 102 | |
| GenBank mRNA | CR406590.1 | - | 2 - 3 | 109 | 102 | |
| M11674.1 | - | 2 - 3 | 112 | 102 | ||
More Information
Set Membership: |
|

LGALS1
Genomic Map