Assay Details
| Gene Symbol: PSMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012543.1 | NP_001012561.1 | 2 - 3 | 259 | 93 | |
| GenBank mRNA | AJ851676.1 | - | 2 - 3 | 259 | 93 | |
| BX931893.1 | - | 1 - 2 | 40 | 93 | ||
| BX950532.1 | - | 1 - 2 | 313 | 93 | ||
More Information
Set Membership: |
|

PSMG1
Genomic Map