Assay Details
| Gene Symbol: RAB11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012551.1 | NP_001012569.1 | 4 - 5 | 573 | 134 | |
| GenBank mRNA | AJ851582.1 | - | 4 - 5 | 573 | 134 | |
More Information
Set Membership: |
|

RAB11B
Genomic Map