Assay Details
| Gene Symbol: ITM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012571.1 | NP_001012589.1 | 3 - 4 | 432 | 66 | |
| GenBank mRNA | AJ851673.1 | - | 3 - 4 | 432 | 66 | |
| CR338976.1 | - | 3 - 4 | 438 | 66 | ||
More Information
Set Membership: |
|

ITM2A
Genomic Map