Assay Details
| Gene Symbol: TMLHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012575.1 | NP_001012593.1 | 2 - 3 | 430 | 61 | |
| XM_015278282.2 | XP_015133768.1 | 2 - 3 | 1434 | 61 | ||
| XM_015278284.2 | XP_015133770.1 | 2 - 3 | 434 | 61 | ||
| XM_025149565.1 | XP_025005333.1 | 2 - 3 | 1126 | 61 | ||
| XM_025149567.1 | XP_025005335.1 | 2 - 3 | 434 | 61 | ||
| GenBank mRNA | AJ851387.1 | - | 2 - 3 | 430 | 61 | |
More Information
Set Membership: |
|

TMLHE
Genomic Map