Assay Details
| Gene Symbol: GNRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012609.1 | NP_001012627.1 | 3 - 4 | 934 | 96 | |
| GenBank mRNA | AY895154.1 | - | 3 - 4 | 934 | 96 | |
| DQ889867.1 | - | 3 - 4 | 1282 | 96 | ||
| DQ889868.1 | - | 3 - 4 | 1087 | 96 | ||
| KJ659046.1 | - | 3 - 4 | 835 | 96 | ||
More Information
Set Membership: |
|

GNRHR
Genomic Map