Assay Details
| Gene Symbol: ASB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012770.1 | NP_001012788.1 | 4 - 5 | 600 | 75 | |
| XM_025153852.1 | XP_025009620.1 | 5 - 6 | 687 | 75 | ||
| GenBank mRNA | AJ851740.1 | - | 4 - 5 | 600 | 75 | |
More Information
Set Membership: |
|

ASB7
Genomic Map