Assay Details
| Gene Symbol: ADAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012779.1 | NP_001012797.1 | 3 - 4 | 786 | 60 | |
| XM_015292606.2 | XP_015148092.1 | 4 - 5 | 920 | 60 | ||
| XM_015292607.2 | XP_015148093.1 | 4 - 5 | 917 | 60 | ||
| XM_015292608.2 | XP_015148094.1 | 3 - 4 | 857 | 60 | ||
| XM_015292609.2 | XP_015148095.1 | 3 - 4 | 851 | 60 | ||
| XM_015292610.2 | XP_015148096.1 | 4 - 5 | 920 | 60 | ||
| XM_025154204.1 | XP_025009972.1 | 3 - 4 | 1448 | 60 | ||
| GenBank mRNA | AJ720968.1 | - | 3 - 4 | 786 | 60 | |
More Information
Set Membership: |
|

ADAT1
Genomic Map