Assay Details
| Gene Symbol: MAP2K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012787.2 | NP_001012805.2 | 9 - 10 | 908 | 61 | |
| XM_025155080.1 | XP_025010848.1 | 9 - 10 | 749 | 61 | ||
| XM_025155081.1 | XP_025010849.1 | 9 - 10 | 793 | 61 | ||
| GenBank mRNA | AJ719928.1 | - | 9 - 10 | 908 | 61 | |
More Information
Set Membership: |
|

MAP2K3
Genomic Map