Assay Details
| Gene Symbol: POLDIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012814.1 | NP_001012832.1 | 6 - 7 | 911 | 73 | |
| XM_015285045.2 | XP_015140531.1 | 6 - 7 | 958 | 73 | ||
| GenBank mRNA | AJ851384.1 | - | 6 - 7 | 911 | 73 | |
| CR353586.1 | - | 5 - 6 | 727 | 73 | ||
More Information
Set Membership: |
|

POLDIP3
Genomic Map