Assay Details
| Gene Symbol: CLDND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012820.1 | NP_001012838.1 | 4 - 5 | 621 | 66 | |
| XM_015295972.2 | XP_015151458.1 | 4 - 5 | 741 | 66 | ||
| XM_015295983.2 | XP_015151469.1 | 3 - 4 | 874 | 66 | ||
| XM_015295985.2 | XP_015151471.1 | 3 - 4 | 864 | 66 | ||
| XM_015295992.2 | XP_015151478.1 | 3 - 4 | 453 | 66 | ||
| GenBank mRNA | AJ720437.1 | - | 4 - 5 | 621 | 66 | |
| CR385544.1 | - | 4 - 5 | 453 | 66 | ||
| CR732891.1 | - | 3 - 4 | 805 | 66 | ||
More Information
Set Membership: |
|

CLDND1
Genomic Map