Assay Details
| Gene Symbol: PSMA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012860.2 | NP_001012878.2 | 4 - 5 | 443 | 73 | |
| GenBank mRNA | AJ721019.1 | - | 3 - 4 | 794 | 73 | |
| CR387901.1 | - | 4 - 5 | 446 | 73 | ||
More Information
Set Membership: |
|

PSMA2
Genomic Map