Assay Details
| Gene Symbol: PELI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012872.2 | NP_001012890.1 | 4 - 5 | 535 | 90 | |
| XM_015278047.2 | XP_015133533.1 | 5 - 6 | 747 | 90 | ||
| XM_015278048.2 | XP_015133534.1 | 5 - 6 | 744 | 90 | ||
| XM_015278049.2 | XP_015133535.1 | 4 - 5 | 586 | 90 | ||
| XM_015278050.2 | XP_015133536.1 | 4 - 5 | 531 | 90 | ||
| XM_015278051.2 | XP_015133537.1 | 4 - 5 | 531 | 90 | ||
| XM_025148641.1 | XP_025004409.1 | 4 - 5 | 391 | 90 | ||
| XM_025148642.1 | XP_025004410.1 | 4 - 5 | 553 | 90 | ||
| GenBank mRNA | AJ719997.1 | - | 4 - 5 | 536 | 90 | |
More Information
Set Membership: |
|

PELI3
Genomic Map