Assay Details
| Gene Symbol: PRIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012880.1 | NP_001012898.1 | 11 - 12 | 1362 | 79 | |
| XM_015284827.2 | XP_015140313.1 | 12 - 13 | 1277 | 79 | ||
| GenBank mRNA | AJ719578.1 | - | 11 - 12 | 1362 | 79 | |
| AJ851645.1 | - | 12 - 13 | 1261 | 79 | ||
More Information
Set Membership: |
|

PRIM2
Genomic Map