Assay Details
| Gene Symbol: RNF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012889.1 | NP_001012907.1 | 7 - 8 | 693 | 84 | |
| XM_015285895.2 | XP_015141381.1 | 6 - 7 | 503 | 84 | ||
| XM_015285896.2 | XP_015141382.1 | 6 - 7 | 775 | 84 | ||
| GenBank mRNA | AJ719403.1 | - | 7 - 8 | 693 | 84 | |
| BX930107.2 | - | 7 - 8 | 671 | 84 | ||
| BX930358.2 | - | 7 - 8 | 682 | 84 | ||
More Information
Set Membership: |
|

RNF4
Genomic Map