Assay Details
| Gene Symbol: PPM1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013394.1 | NP_001013412.1 | 7 - 8 | 985 | 76 | |
| GenBank mRNA | AJ719435.1 | - | 7 - 8 | 985 | 76 | |
| CR523416.1 | - | 6 - 7 | 690 | 76 | ||
More Information
Set Membership: |
|

PPM1M
Genomic Map