Assay Details
| Gene Symbol: SMAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014968.1 | NP_001014968.1 | 4 - 5 | 1014 | 92 | |
| XM_015293967.2 | XP_015149453.1 | 5 - 6 | 1324 | 92 | ||
| XM_015293968.2 | XP_015149454.1 | 5 - 6 | 1246 | 92 | ||
| XM_025154805.1 | XP_025010573.1 | 6 - 7 | 4259 | 92 | ||
| GenBank mRNA | AF143240.1 | - | 4 - 5 | 901 | 92 | |
| AY953144.1 | - | 4 - 5 | 1014 | 92 | ||
More Information
Set Membership: |
|

SMAD5
Genomic Map