Assay Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029981.2 | NP_001025152.1 | 3 - 4 | 267 | 87 | |
| XM_015296367.2 | XP_015151853.1 | 3 - 4 | 317 | 87 | ||
| XM_015296368.2 | XP_015151854.1 | 3 - 4 | 261 | 87 | ||
| XM_015296369.2 | XP_015151855.1 | 3 - 4 | 289 | 87 | ||
| XM_015296370.2 | XP_015151856.1 | 3 - 4 | 283 | 87 | ||
| XM_015296371.2 | XP_015151857.1 | 3 - 4 | 1180 | 87 | ||
| XM_015296372.2 | XP_015151858.1 | 3 - 4 | 1189 | 87 | ||
| XM_025142197.1 | XP_024997965.1 | 3 - 4 | 260 | 87 | ||
| XM_025142198.1 | XP_024997966.1 | 3 - 4 | 236 | 87 | ||
| XM_025142199.1 | XP_024997967.1 | 3 - 4 | 249 | 87 | ||
| XM_025142200.1 | XP_024997968.1 | 3 - 4 | 224 | 87 | ||
| GenBank mRNA | BX930492.2 | - | 3 - 4 | 267 | 87 | |
| BX931834.1 | - | 3 - 4 | 270 | 87 | ||
| BX935154.1 | - | 3 - 4 | 268 | 87 | ||
More Information
Set Membership: |
|

OSER1
Genomic Map