Assay Details
| Gene Symbol: EEF2KMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030627.1 | NP_001025798.1 | 7 - 8 | 1059 | 71 | |
| GenBank mRNA | AJ720764.1 | - | 7 - 8 | 1059 | 71 | |
| CR524108.1 | - | 5 - 6 | 892 | 71 | ||
More Information
Set Membership: |
|

EEF2KMT
Genomic Map