Assay Details
| Gene Symbol: CYP2W1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642141.3 | XP_003642189.2 | 3 - 4 | 608 | 95 | |
| XM_015294328.2 | XP_015149814.1 | 3 - 4 | 1461 | 95 | ||
| XM_025155214.1 | XP_025010982.1 | 3 - 4 | 415 | 95 | ||
| XM_025155215.1 | XP_025010983.1 | 3 - 4 | 320 | 95 | ||
| GenBank mRNA | AJ720136.1 | - | 609 | 95 | ||
More Information
Set Membership: |
|

CYP2W1
Genomic Map