Assay Details
| Gene Symbol: ARPC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030632.1 | NP_001025803.1 | 6 - 7 | 852 | 71 | |
| XM_015294172.2 | XP_015149658.1 | 6 - 7 | 885 | 71 | ||
| GenBank mRNA | AJ720885.1 | - | 6 - 7 | 852 | 71 | |
More Information
Set Membership: |
|

ARPC1B
Genomic Map