Assay Details
| Gene Symbol: HAGHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030637.1 | NP_001025808.1 | 1 - 2 | 198 | 99 | |
| GenBank mRNA | AJ719602.1 | - | 1 - 2 | 198 | 99 | |
| AJ851726.1 | - | 1 - 2 | 160 | 99 | ||
More Information
Set Membership: |
|

HAGHL
Genomic Map