Assay Details
| Gene Symbol: PPP1R16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030851.1 | NP_001026022.1 | 4 - 5 | 1019 | 66 | |
| XM_015296355.2 | XP_015151841.1 | 4 - 5 | 1186 | 66 | ||
| XM_015296357.2 | XP_015151843.1 | 4 - 5 | 1740 | 66 | ||
| GenBank mRNA | AJ721001.1 | - | 4 - 5 | 1019 | 66 | |
More Information
Set Membership: |
|

PPP1R16B
Genomic Map