Assay Details
| Gene Symbol: GTF3C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031076.1 | NP_001026247.1 | 1 - 2 | 132 | 60 | |
| XM_015284508.2 | XP_015139994.1 | 1 - 2 | 254 | 60 | ||
| GenBank mRNA | AJ720304.1 | - | 1 - 2 | 132 | 60 | |
More Information
Set Membership: |
|

GTF3C6
Genomic Map