Assay Details
| Gene Symbol: GTF3C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031076.1 | NP_001026247.1 | 5 - 6 | 438 | 108 | |
| XM_015284508.2 | XP_015139994.1 | 5 - 6 | 563 | 108 | ||
| XM_015284509.2 | XP_015139995.1 | 5 - 6 | 457 | 108 | ||
| XM_015284510.2 | XP_015139996.1 | 5 - 6 | 699 | 108 | ||
| XM_015284511.2 | XP_015139997.1 | 5 - 6 | 689 | 108 | ||
| GenBank mRNA | AJ720304.1 | - | 5 - 6 | 438 | 108 | |
More Information
Set Membership: |
|

GTF3C6
Genomic Map