Assay Details
| Gene Symbol: PSMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031190.1 | NP_001026361.1 | 5 - 6 | 605 | 80 | |
| XM_025150705.1 | XP_025006473.1 | 5 - 6 | 437 | 80 | ||
| GenBank mRNA | AJ720697.1 | - | 5 - 6 | 605 | 80 | |
| CR390353.1 | - | 5 - 6 | 408 | 80 | ||
More Information
Set Membership: |
|

PSMC3
Genomic Map