Assay Details
| Gene Symbol: CCNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031209.1 | NP_001026380.1 | 8 - 9 | 1208 | 72 | |
| XM_015287468.2 | XP_015142954.1 | 7 - 8 | 1695 | 72 | ||
| GenBank mRNA | AJ719906.1 | - | 8 - 9 | 1208 | 72 | |
More Information
Set Membership: |
|

CCNK
Genomic Map