Assay Details
| Gene Symbol: YY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031210.1 | NP_001026381.1 | 3 - 4 | 1200 | 66 | |
| GenBank mRNA | AJ719349.1 | - | 3 - 4 | 1200 | 66 | |
More Information
Set Membership: |
|

YY2
Genomic Map