Assay Details
| Gene Symbol: GMFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031214.1 | NP_001026385.1 | 2 - 3 | 158 | 78 | |
| GenBank mRNA | AJ720612.1 | - | 2 - 3 | 158 | 78 | |
More Information
Set Membership: |
|

GMFB
Genomic Map