Assay Details
| Gene Symbol: RGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031216.1 | NP_001026387.1 | 1 - 2 | 150 | 57 | |
| GenBank mRNA | AY339627.1 | - | 1 - 2 | 150 | 57 | |
More Information
Set Membership: |
|

RGR
Genomic Map