Assay Details
| Gene Symbol: ARID5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031220.1 | NP_001026391.1 | 8 - 9 | 1207 | 105 | |
| XM_015288120.2 | XP_015143606.1 | 7 - 8 | 1265 | 105 | ||
| XM_015288121.2 | XP_015143607.1 | 5 - 6 | 1007 | 105 | ||
| XM_015288128.2 | XP_015143614.1 | 15 - 16 | 5524 | 105 | ||
| GenBank mRNA | AJ720565.1 | - | 8 - 9 | 1207 | 105 | |
More Information
Set Membership: |
|

ARID5B
Genomic Map