Assay Details
| Gene Symbol: SGTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031418.1 | NP_001026589.1 | 10 - 11 | 886 | 68 | |
| GenBank mRNA | AJ720539.1 | - | 10 - 11 | 886 | 68 | |
More Information
Set Membership: |
|

SGTB
Genomic Map