Assay Details
| Gene Symbol: CYP26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001129.1 | NP_001001129.1 | 4 - 5 | 843 | 121 | |
| XM_015288598.2 | XP_015144084.1 | 5 - 6 | 758 | 121 | ||
| GenBank mRNA | AF199462.1 | - | 4 - 5 | 843 | 121 | |
More Information
Set Membership: |
|

CYP26A1
Genomic Map