Assay Details
| Gene Symbol: HSD11B1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001201.1 | NP_001001201.1 | 2 - 3 | 204 | 116 | |
| XM_015299681.2 | XP_015155167.2 | 1 - 2 | 462 | 116 | ||
| XM_015299683.2 | XP_015155169.2 | 2 - 3 | 304 | 116 | ||
| XM_015299684.2 | XP_015155170.2 | 2 - 3 | 300 | 116 | ||
| XM_015299685.2 | XP_015155171.2 | 2 - 3 | 354 | 116 | ||
| XM_015299686.2 | XP_015155172.2 | 2 - 3 | 224 | 116 | ||
| XM_015299687.2 | XP_015155173.2 | 2 - 3 | 178 | 116 | ||
| XM_015299688.1 | XP_015155174.1 | 1 - 2 | 462 | 116 | ||
| XM_015299689.2 | XP_015155175.1 | 1 - 2 | 462 | 116 | ||
| XM_015299690.2 | XP_015155176.1 | 1 - 2 | 462 | 116 | ||
| XM_015299691.2 | XP_015155177.2 | 1 - 2 | 462 | 116 | ||
| XM_015299692.1 | XP_015155178.1 | 1 - 2 | 462 | 116 | ||
| XM_015299693.2 | XP_015155179.2 | 2 - 3 | 297 | 116 | ||
| XM_025144260.1 | XP_025000028.1 | 1 - 2 | 462 | 116 | ||
| XM_025144261.1 | XP_025000029.1 | 2 - 3 | 236 | 116 | ||
| GenBank mRNA | AY578179.1 | - | 2 - 3 | 204 | 116 | |
| CR406217.1 | - | 2 - 3 | 194 | 116 | ||
More Information
Set Membership: |
|

HSD11B1L
Genomic Map