Assay Details
| Gene Symbol: RTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001466.2 | NP_001001466.1 | 8 - 9 | 2373 | 63 | |
| XM_015287270.2 | XP_015142756.1 | 6 - 7 | 781 | 63 | ||
| XM_025150542.1 | XP_025006310.1 | 9 - 10 | 2224 | 63 | ||
| GenBank mRNA | AY164723.1 | - | 6 - 7 | 759 | 63 | |
| AY164724.1 | - | 8 - 9 | 2374 | 63 | ||
| U17605.1 | - | 6 - 7 | 759 | 63 | ||
| U17606.1 | - | 8 - 9 | 2374 | 63 | ||
More Information
Set Membership: |
|

RTN1
Genomic Map