Assay Details
| Gene Symbol: ST8SIA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001604.1 | NP_001001604.1 | 3 - 4 | 293 | 70 | |
| XM_015292082.2 | XP_015147568.1 | 2 - 3 | 334 | 70 | ||
| GenBank mRNA | AJ699419.1 | - | 3 - 4 | 293 | 70 | |
| CR406140.1 | - | 2 - 3 | 288 | 70 | ||
More Information
Set Membership: |
|

ST8SIA2
Genomic Map