Assay Details
| Gene Symbol: UBA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001765.1 | NP_001001765.1 | 4 - 5 | 379 | 75 | |
| XM_025147083.1 | XP_025002851.1 | 2 - 3 | 188 | 75 | ||
| GenBank mRNA | AY620963.1 | - | 4 - 5 | 379 | 75 | |
| CR387232.1 | - | 2 - 3 | 161 | 75 | ||
More Information
Set Membership: |
|

UBA5
Genomic Map