Assay Details
| Gene Symbol: TCP11X2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001897.1 | NP_001001897.1 | 5 - 6 | 770 | 120 | |
| XM_015285915.2 | XP_015141401.1 | 6 - 7 | 865 | 120 | ||
| XM_015285922.2 | XP_015141408.1 | 6 - 7 | 862 | 120 | ||
| XM_015285929.2 | XP_015141415.1 | 6 - 7 | 1043 | 120 | ||
| XM_025150870.1 | XP_025006638.1 | 4 - 5 | 612 | 120 | ||
| GenBank mRNA | AJ851377.1 | - | 6 - 7 | 822 | 120 | |
| AY633747.1 | - | 5 - 6 | 770 | 120 | ||
More Information
Set Membership: |
|

TCP11X2
Genomic Map