Assay Details
| Gene Symbol: TCP11X2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001897.1 | NP_001001897.1 | 6 - 7 | 904 | 143 | |
| XM_015285915.2 | XP_015141401.1 | 7 - 8 | 999 | 143 | ||
| XM_015285922.2 | XP_015141408.1 | 7 - 8 | 996 | 143 | ||
| XM_015285929.2 | XP_015141415.1 | 7 - 8 | 1177 | 143 | ||
| XM_025150870.1 | XP_025006638.1 | 5 - 6 | 746 | 143 | ||
| GenBank mRNA | AJ851377.1 | - | 7 - 8 | 956 | 143 | |
| AY633747.1 | - | 6 - 7 | 904 | 143 | ||
More Information
Set Membership: |
|

TCP11X2
Genomic Map