Assay Details
| Gene Symbol: CFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004406.1 | NP_001004406.1 | 3 - 4 | 427 | 99 | |
| XM_015287368.2 | XP_015142854.1 | 3 - 4 | 713 | 99 | ||
| XM_015287369.2 | XP_015142855.1 | 3 - 4 | 516 | 99 | ||
| GenBank mRNA | CR354089.1 | - | 3 - 4 | 338 | 99 | |
| M55659.1 | - | 3 - 4 | 427 | 99 | ||
More Information
Set Membership: |
|

CFL2
Genomic Map