Assay Details
| Gene Symbol: JCHAIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204263.1 | NP_989594.1 | 3 - 4 | 412 | 72 | |
| GenBank mRNA | AB025103.1 | - | 3 - 4 | 412 | 72 | |
More Information
Set Membership: |
|

JCHAIN
Genomic Map