Assay Details
| Gene Symbol: GABRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204318.2 | NP_989649.1 | 5 - 6 | 723 | 68 | |
| XM_015293637.2 | XP_015149123.1 | 5 - 6 | 606 | 68 | ||
| XM_025154781.1 | XP_025010549.1 | 5 - 6 | 652 | 68 | ||
| GenBank mRNA | X54244.1 | - | 664 | 68 | ||
More Information
Set Membership: |
|

GABRA1
Genomic Map