Assay Details
| Gene Symbol: BID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204552.2 | NP_989883.2 | 5 - 6 | 670 | 64 | |
| XM_015284410.2 | XP_015139896.1 | 5 - 6 | 851 | 64 | ||
| XM_025153522.1 | XP_025009290.1 | 7 - 8 | 1148 | 64 | ||
| XM_025153546.1 | XP_025009314.1 | 5 - 6 | 803 | 64 | ||
| GenBank mRNA | AJ719847.1 | - | 5 - 6 | 670 | 64 | |
| AY112660.1 | - | 5 - 6 | 700 | 64 | ||
| CR387911.1 | - | 1 - 2 | 320 | 64 | ||
More Information
Set Membership: |
|

BID
Genomic Map