Assay Details
| Gene Symbol: CD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204921.1 | NP_990252.1 | 5 - 6 | 764 | 115 | |
| GenBank mRNA | Y12011.1 | - | 764 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD5| Gene Symbol: CD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204921.1 | NP_990252.1 | 5 - 6 | 764 | 115 | |
| GenBank mRNA | Y12011.1 | - | 764 | 115 | ||
Set Membership: |
|